Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125994.2 | NP_201398.1 | 11 - 12 | 1493 | 78 | |
GenBank mRNA | AF369571.1 | - | 11 - 12 | 1178 | 78 | |
BT033095.1 | - | 11 - 12 | 1133 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |