Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SD1-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101008.3 | NP_172602.1 | 2 - 3 | 1526 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |