Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101060.4 | NP_172653.1 | 3 - 4 | 521 | 70 | |
GenBank mRNA | AY045795.1 | - | 3 - 4 | 469 | 70 | |
AY079413.1 | - | 2 - 3 | 352 | 70 | ||
AY086749.1 | - | 3 - 4 | 499 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |