Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATBETAFRUCT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101096.3 | NP_563901.1 | 4 - 5 | 1332 | 86 | |
GenBank mRNA | AY046009.1 | - | 4 - 5 | 1313 | 86 | |
AY142666.1 | - | 4 - 5 | 1274 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |