Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101141.4 | NP_172731.1 | 2 - 3 | 738 | 136 | |
GenBank mRNA | AK117662.1 | - | 2 - 3 | 603 | 136 | |
BT005109.1 | - | 2 - 3 | 446 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |