Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP87A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198045.1 | NP_001184974.1 | 8 - 9 | 1521 | 70 | |
NM_001332045.1 | NP_001320967.1 | 7 - 8 | 1598 | 70 | ||
NM_001332046.1 | NP_001320968.1 | 8 - 9 | 1521 | 70 | ||
NM_101144.5 | NP_172734.2 | 8 - 9 | 1576 | 70 | ||
GenBank mRNA | AK176324.1 | - | 8 - 9 | 1501 | 70 | |
BT010888.1 | - | 8 - 9 | 1323 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |