Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084060.1 | NP_001077529.1 | 1 - 2 | 158 | 75 | |
NM_001198048.1 | NP_001184977.1 | 1 - 2 | 152 | 75 | ||
NM_001332055.1 | NP_001321059.1 | 1 - 2 | 212 | 75 | ||
NM_101161.3 | NP_172750.1 | 1 - 2 | 158 | 75 | ||
GenBank mRNA | BT008637.1 | - | 1 - 2 | 69 | 75 | |
BT015867.1 | - | 1 - 2 | 41 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |