Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198051.1 | NP_001184980.1 | 1 - 2 | 862 | 65 | |
NM_101187.3 | NP_172776.1 | 1 - 2 | 942 | 65 | ||
GenBank mRNA | AF462814.1 | - | 1 - 2 | 858 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |