Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101188.3 | NP_172777.1 | 6 - 7 | 1453 | 93 | |
GenBank mRNA | AY093149.1 | - | 6 - 7 | 1349 | 93 | |
BT006584.1 | - | 6 - 7 | 983 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |