Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101266.4 | NP_172853.1 | 8 - 9 | 1230 | 154 | |
GenBank mRNA | BT026491.1 | - | 8 - 9 | 982 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |