Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101295.5 | NP_563944.1 | 1 - 2 | 700 | 117 | |
GenBank mRNA | AF361856.1 | - | 1 - 2 | 387 | 117 | |
BT001128.1 | - | 1 - 2 | 219 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |