Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101308.4 | NP_172893.1 | 1 - 2 | 493 | 68 | |
GenBank mRNA | AF332452.1 | - | 1 - 2 | 223 | 68 | |
AF370156.1 | - | 1 - 2 | 306 | 68 | ||
AY059097.1 | - | 1 - 2 | 223 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |