Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101308.4 | NP_172893.1 | 2 - 3 | 591 | 83 | |
GenBank mRNA | AF332452.1 | - | 2 - 3 | 321 | 83 | |
AF370156.1 | - | 2 - 3 | 404 | 83 | ||
AY059097.1 | - | 2 - 3 | 321 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |