Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160865.1 | NP_001154337.1 | 7 - 8 | 951 | 82 | |
NM_001332123.1 | NP_001320937.1 | 7 - 8 | 1261 | 82 | ||
NM_101319.4 | NP_172904.2 | 8 - 9 | 1161 | 82 | ||
GenBank mRNA | AK175115.1 | - | 7 - 8 | 1023 | 82 | |
AK176690.1 | - | 7 - 8 | 1023 | 82 | ||
AY136388.1 | - | 7 - 8 | 964 | 82 | ||
BT000187.1 | - | 7 - 8 | 904 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |