Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101348.4 | NP_172932.1 | 3 - 4 | 2877 | 113 | |
GenBank mRNA | AK228731.1 | - | 3 - 4 | 2621 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |