Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101352.3 | NP_172936.1 | 12 - 13 | 1566 | 98 | |
GenBank mRNA | AJ304841.1 | - | 12 - 13 | 1490 | 98 | |
AY039955.1 | - | 12 - 13 | 1451 | 98 | ||
AY150494.1 | - | 12 - 13 | 1290 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |