Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101357.4 | NP_172941.1 | 2 - 3 | 359 | 96 | |
GenBank mRNA | AK118861.1 | - | 2 - 3 | 200 | 96 | |
BT005663.1 | - | 2 - 3 | 162 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |