Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106129.4 | NP_565090.1 | 7 - 8 | 1309 | 125 | |
NM_202414.1 | NP_974143.1 | 7 - 8 | 1308 | 125 | ||
GenBank mRNA | AF367342.1 | - | 7 - 8 | 1307 | 125 | |
AY056055.1 | - | 7 - 8 | 1309 | 125 | ||
AY124864.1 | - | 7 - 8 | 1213 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |