Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106132.4 | NP_177612.2 | 3 - 4 | 1131 | 87 | |
GenBank mRNA | AY140016.1 | - | 3 - 4 | 1104 | 87 | |
BT008900.1 | - | 3 - 4 | 936 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |