Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NdhO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106146.5 | NP_565093.1 | 5 - 6 | 505 | 82 | |
GenBank mRNA | AF410289.1 | - | 5 - 6 | 505 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |