Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106147.2 | NP_177627.1 | 3 - 4 | 591 | 68 | |
GenBank mRNA | AF305720.1 | - | 3 - 4 | 276 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |