Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH10A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198470.1 | NP_001185399.1 | 2 - 3 | 402 | 71 | |
NM_106150.4 | NP_565094.1 | 2 - 3 | 510 | 71 | ||
GenBank mRNA | AY087395.1 | - | 2 - 3 | 417 | 71 | |
AY093071.1 | - | 2 - 3 | 375 | 71 | ||
BT008872.1 | - | 2 - 3 | 246 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |