Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106166.4 | NP_565101.1 | 3 - 4 | 1732 | 106 | |
GenBank mRNA | AB158477.1 | - | 3 - 4 | 1446 | 106 | |
AY057504.1 | - | 3 - 4 | 1464 | 106 | ||
AY103303.1 | - | 3 - 4 | 1433 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |