Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106166.4 | NP_565101.1 | 7 - 8 | 2033 | 110 | |
GenBank mRNA | AB158477.1 | - | 7 - 8 | 1747 | 110 | |
AY057504.1 | - | 7 - 8 | 1765 | 110 | ||
AY103303.1 | - | 7 - 8 | 1734 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |