Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084358.2 | NP_001077827.1 | 3 - 4 | 1862 | 72 | |
NM_106195.5 | NP_177674.1 | 4 - 5 | 1567 | 72 | ||
GenBank mRNA | AF353093.1 | - | 3 - 4 | 1214 | 72 | |
AY085278.1 | - | 3 - 4 | 1208 | 72 | ||
AY598452.1 | - | 4 - 5 | 1328 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |