Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106199.5 | NP_177678.2 | 1 - 2 | 900 | 99 | |
GenBank mRNA | AK176378.1 | - | 1 - 2 | 900 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |