Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106219.3 | NP_177697.1 | 3 - 4 | 1342 | 113 | |
GenBank mRNA | AY039938.1 | - | 3 - 4 | 1238 | 113 | |
AY150451.1 | - | 3 - 4 | 938 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |