Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334692.1 | NP_001321036.1 | 4 - 5 | 429 | 80 | |
NM_001334693.1 | NP_001321035.1 | 3 - 4 | 413 | 80 | ||
NM_106221.5 | NP_177699.4 | 3 - 4 | 247 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |