Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106229.2 | NP_177707.1 | 3 - 4 | 658 | 105 | |
GenBank mRNA | DQ446433.1 | - | 3 - 4 | 518 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |