Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106241.3 | NP_177719.1 | 4 - 5 | 944 | 133 | |
GenBank mRNA | AK227028.1 | - | 4 - 5 | 942 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |