Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SmD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106280.4 | NP_177757.1 | 2 - 3 | 293 | 88 | |
GenBank mRNA | AF380651.1 | - | 2 - 3 | 222 | 88 | |
AY054149.1 | - | 2 - 3 | 207 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |