Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334740.1 | NP_001321484.1 | 9 - 10 | 895 | 120 | |
NM_001334741.1 | NP_001321485.1 | 9 - 10 | 860 | 120 | ||
NM_106321.4 | NP_177797.2 | 9 - 10 | 878 | 120 | ||
NM_202429.2 | NP_974158.1 | 8 - 9 | 1007 | 120 | ||
GenBank mRNA | AB493538.1 | - | 8 - 9 | 738 | 120 | |
BT001913.1 | - | 8 - 9 | 738 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |