Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106362.3 | NP_177837.1 | 2 - 3 | 426 | 69 | |
GenBank mRNA | AY045612.1 | - | 2 - 3 | 227 | 69 | |
AY090330.1 | - | 2 - 3 | 172 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |