Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106387.3 | NP_177862.1 | 4 - 5 | 775 | 60 | |
GenBank mRNA | AY099788.1 | - | 4 - 5 | 775 | 60 | |
AY128905.1 | - | 4 - 5 | 637 | 60 | ||
X77499.1 | - | 724 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |