Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106425.3 | NP_177899.1 | 2 - 3 | 1309 | 75 | |
GenBank mRNA | AF424624.1 | - | 2 - 3 | 1268 | 75 | |
AY090950.1 | - | 2 - 3 | 1270 | 75 | ||
BT000989.1 | - | 2 - 3 | 1192 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |