Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106425.3 | NP_177899.1 | 3 - 4 | 1540 | 89 | |
GenBank mRNA | AF424624.1 | - | 3 - 4 | 1499 | 89 | |
AY090950.1 | - | 3 - 4 | 1501 | 89 | ||
BT000989.1 | - | 3 - 4 | 1423 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |