Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106434.2 | NP_565161.1 | 2 - 3 | 2082 | 65 | |
GenBank mRNA | BT002749.1 | - | 2 - 3 | 1751 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |