Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106463.3 | NP_177937.1 | 1 - 2 | 319 | 66 | |
GenBank mRNA | AK227331.1 | - | 1309 | 66 | ||
BT004567.1 | - | 1 - 2 | 288 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |