Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161003.2 | NP_001154475.1 | 9 - 10 | 2331 | 108 | |
NM_106474.5 | NP_177948.3 | 8 - 9 | 2446 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |