Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198498.1 | NP_001185427.1 | 8 - 9 | 1617 | 88 | |
NM_001334830.1 | NP_001322353.1 | 8 - 9 | 1715 | 88 | ||
NM_106501.5 | NP_177975.3 | 8 - 9 | 1715 | 88 | ||
GenBank mRNA | AK221532.1 | - | 8 - 9 | 1605 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |