Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334832.1 | NP_001319403.1 | 15 - 16 | 4099 | 70 | |
NM_001334833.1 | NP_001322931.1 | 14 - 15 | 4058 | 70 | ||
NM_001334834.1 | NP_001322932.1 | 14 - 15 | 4025 | 70 | ||
NM_106505.5 | NP_177979.1 | 14 - 15 | 2289 | 70 | ||
GenBank mRNA | KJ138926.1 | - | 13 - 14 | 2083 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |