Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334835.1 | NP_001321203.1 | 1 - 2 | 187 | 68 | |
NM_106506.3 | NP_177980.2 | 1 - 2 | 195 | 68 | ||
GenBank mRNA | BT022078.1 | - | 1 - 2 | 117 | 68 | |
BT033136.1 | - | 1 - 2 | 93 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |