Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124147.1 | NP_001117619.1 | 2 - 3 | 547 | 99 | |
NM_001334854.1 | NP_001322698.1 | 2 - 3 | 562 | 99 | ||
NM_106541.4 | NP_565195.1 | 2 - 3 | 629 | 99 | ||
GenBank mRNA | AB034696.1 | - | 2 - 3 | 467 | 99 | |
AY054485.1 | - | 2 - 3 | 547 | 99 | ||
BT010353.1 | - | 1 - 2 | 96 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |