Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334861.1 | NP_001322959.1 | 3 - 4 | 804 | 109 | |
NM_001334862.1 | NP_001322960.1 | 4 - 5 | 570 | 109 | ||
NM_001334863.1 | NP_001322961.1 | 3 - 4 | 728 | 109 | ||
NM_001334864.1 | NP_001322958.1 | 2 - 3 | 298 | 109 | ||
NM_106545.4 | NP_178017.2 | 4 - 5 | 599 | 109 | ||
GenBank mRNA | AY091131.1 | - | 4 - 5 | 523 | 109 | |
AY114039.1 | - | 3 - 4 | 476 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |