Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106547.3 | NP_178019.2 | 4 - 5 | 509 | 64 | |
GenBank mRNA | AY518290.1 | - | 4 - 5 | 509 | 64 | |
FJ708683.1 | - | 4 - 5 | 354 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |