Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334878.1 | NP_001321640.1 | 3 - 4 | 1205 | 114 | |
NM_001334879.1 | NP_001321639.1 | 3 - 4 | 1200 | 114 | ||
NM_001334880.1 | NP_001321641.1 | 3 - 4 | 563 | 114 | ||
NM_106577.4 | NP_565205.1 | 3 - 4 | 916 | 114 | ||
GenBank mRNA | AF326910.1 | - | 3 - 4 | 510 | 114 | |
AY113874.1 | - | 3 - 4 | 229 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |