Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106579.5 | NP_178049.2 | 1 - 2 | 492 | 83 | |
GenBank mRNA | AY219832.1 | - | 1 - 2 | 345 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |