Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPCP-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106601.4 | NP_178071.1 | 1 - 2 | 418 | 60 | |
GenBank mRNA | AF348583.1 | - | 1 - 2 | 98 | 60 | |
AK117920.1 | - | 1 - 2 | 244 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |