Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPCP-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106601.4 | NP_178071.1 | 6 - 7 | 804 | 65 | |
GenBank mRNA | AF348583.1 | - | 6 - 7 | 484 | 65 | |
AK117920.1 | - | 6 - 7 | 630 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |