Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124150.1 | NP_001117622.1 | 19 - 20 | 3026 | 74 | |
NM_001198512.1 | NP_001185441.1 | 19 - 20 | 3199 | 74 | ||
NM_001198513.2 | NP_001185442.1 | 19 - 20 | 3139 | 74 | ||
NM_106632.4 | NP_178101.3 | 19 - 20 | 3126 | 74 | ||
GenBank mRNA | AK226727.1 | - | 19 - 20 | 3028 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |